Learn More
Medchemexpress LLC HY-B0686 10mg Medchemexpress, Eptifibatide CAS:188627-80-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB068610MG
Encompass_Preferred
Medchemexpress, HY-B0686 10mg Eptifibatide CAS:188627-80-7 Eptifibatide is a cyclic heptapeptide, acts as a competitive antagonist for the activated platelet glycoprotein IIb/IIIa receptor, with anti-platelet activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2641