Learn More
Apexbio Technology LLC Eptifibatide 188627-80-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063781
Encompass_Preferred
Eptifibatide is a cyclic heptapeptide belonging to the glycoprotein IIb/IIIa inhibitor category functioning as an antagonist of platelet adhesion by reversibly binding to the platelet glycoprotein IIb/IIIa receptor This receptor located on platelet surfaces mediates platelet aggregation through binding fibrinogen and von Willebrand factor processes essential in thrombus formation By blocking the GP IIb/IIIa receptor binding site Eptifibatide inhibits fibrinogen-dependent platelet cross-linking reducing platelet aggregation In biomedical research Eptifibatide serves as a pharmacological tool to investigate platelet function thrombosis mechanisms and related cardiovascular diseases in preclinical and clinical models
Catalog No. 50-000-63781 Supplier Apexbio Technology LLC Supplier No. B34905.1
Eptifibatide is a cyclic heptapeptide belonging to the glycoprotein IIb/IIIa inhibitor category functioning as an antagonist of platelet adhesion by reversibly binding to the platelet glycoprotein IIb/IIIa receptor This receptor located on platelet surfaces mediates platelet aggregation through binding fibrinogen and von Willebrand factor processes essential in thrombus formation By blocking the GP IIb/IIIa receptor binding site Eptifibatide inhibits fibrinogen-dependent platelet cross-linking reducing platelet aggregation In biomedical research Eptifibatide serves as a pharmacological tool to investigate platelet function thrombosis mechanisms and related cardiovascular diseases in preclinical and clinical models