Learn More
TARGETMOL CHEMICALS INC EPTIFIBATIDE ACETATE 148 50MG
SDP
Catalog No. 502579804
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Eptifibatide acetate (148031-34-9 free base) (Eptifibatide acetate) an antiplatelet drug of the glycoprotein IIb/IIIa inhibitor class is a cyclic heptapeptide constructed from 6 amino acids and a mercaptopropionyl residue. purity: 99%
Catalog No. 50-257-9804 Supplier TARGETMOL CHEMICALS INC Supplier No. T514650MG
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Eptifibatide acetate (148031-34-9 free base) (Eptifibatide acetate) an antiplatelet drug of the glycoprotein IIb/IIIa inhibitor class is a cyclic heptapeptide constructed from 6 amino acids and a mercaptopropionyl residue. purity: 99%