Learn More
Selleck Chemical LLC Eptifibatide Acetate 50mg 148031-34-9 Mpr-Har-Gly-Asp-Trp-Pro-Cys-NH2
SDP
Catalog No. 501948181
Encompass_Preferred
Eptifibatide Acetate is a cyclic heptapeptide constructed from 6 amino acids and a mercaptopropionyl residue. A disulfide bridge forms between mercaptopropionyl and cysteine. Eptifibatide prevents platelet aggregation. One study shows the inhibition of platelet glycoprotein IIb/IIIa in acute coronary syndromes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8181 Supplier Selleck Chemical LLC Supplier No. P101150MG
Eptifibatide Acetate is a cyclic heptapeptide constructed from 6 amino acids and a mercaptopropionyl residue. A disulfide bridge forms between mercaptopropionyl and cysteine. Eptifibatide prevents platelet aggregation. One study shows the inhibition of platelet glycoprotein IIb/IIIa in acute coronary syndromes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.