Learn More
Medchemexpress LLC Eptifibatide acetate | 881997-86-0 | 99.67% | 5 MG
SDP
Catalog No. 5000408266
Encompass_Preferred
Eptifibatide acetate is a cyclic heptapeptide that functions as a competitive antagonist for the activated platelet glycoprotein IIb/IIIa receptor, exhibiting anti-platelet activity. It is intended for research use only.
  • Cyclic heptapeptide
  • Competitive antagonist for activated platelet glycoprotein IIb/IIIa receptor
  • Anti-platelet activity
Catalog No. 50-004-08266 Supplier Medchemexpress LLC Supplier No. HYB0686B5MG
Eptifibatide acetate is a cyclic heptapeptide that functions as a competitive antagonist for the activated platelet glycoprotein IIb/IIIa receptor, exhibiting anti-platelet activity. It is intended for research use only.
  • Cyclic heptapeptide
  • Competitive antagonist for activated platelet glycoprotein IIb/IIIa receptor
  • Anti-platelet activity