Learn More
Cayman Chemical EptIfIbatIdeacetate 50mg
SDP
Catalog No. 502964221
Encompass_Preferred
A potent GPIIb/IIIa antagonist (Kd = 120 nM); prevents binding of adhesive proteins fibrinogen and von Willebrand factor to GPIIb/IIIa on the surface of activated platelets to prevent aggregation and thrombus formation; inhibits ADP-induced citrated blood aggregation (IC50 = 0.11-0.22 μg/ml) in vitro and in vivo (IC50 = 52 μg/ml in porcine plasma)
Catalog No. 50-296-4221 Supplier Cayman Chemical Supplier No. 2157850 MG
A potent GPIIb/IIIa antagonist (Kd = 120 nM); prevents binding of adhesive proteins fibrinogen and von Willebrand factor to GPIIb/IIIa on the surface of activated platelets to prevent aggregation and thrombus formation; inhibits ADP-induced citrated blood aggregation (IC50 = 0.11-0.22 μg/ml) in vitro and in vivo (IC50 = 52 μg/ml in porcine plasma)