Learn More
Biomatik Corporation Epx Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11373
Encompass_Preferred
Epx Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to EPX. The Epx Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Eosinophil Peroxidase Protein (140-300Aa). Applications: ELISA, IHC. Target Synonyms: EPX. Accession Number: P11678. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5371