Learn More
Selleck Chemical LLC Tazemetostat (EPZ-6438) 10mg 1403254-99-8 E7438
SDP
Supplier:  Selleck Chemical LLC S712810MG
Encompass_Preferred
Tazemetostat (EPZ-6438, E7438) is a potent, and selective EZH2 inhibitor with Ki and IC50 of 2.5 nM and 11 nM in cell-free assays, exhibiting a 35-fold selectivity versus EZH1 and >4,500-fold selectivity relative to 14 other HMTs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0798314