Learn More
Medchemexpress LLC HY-13803 200mg , EPZ-6438 Tazemetostat;E-7438 CAS:1403254-99-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13803200MG
Encompass_Preferred
Medchemexpress, HY-13803 200mg EPZ-6438 Tazemetostat;E-7438 CAS:1403254-99-8 Formula:C34H44N4O4 Ki: 2.5 nM (EZH2) Purity:98% EPZ-6438 inhibits the activity of human PRC2-containing wild-type EZH2 with K i of 2.5±0.5 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2521