Learn More
Apexbio Technology LLC EPZ-6438, 50mg. Cas: 1403254-99-8 MFCD: MFCD24849415. EZH2 inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC A822150
Encompass_Preferred
EPZ-6438 is a potent and bio-available inhibitor of EZH2, the catalytic subunit of polycomb repressive complex 2 (PRC2) catalyzing the methylation of lysine 27 of histone H3 (H3K27), that inhibits the activity of human PRC2-containing wild-type EZH2 with a value of inhibition constant Ki of 2.5 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5081