Learn More
Selleck Chemical LLC EPZ004777 5mg 1338466-77-5
SDP
Supplier:  Selleck Chemical LLC S73535MG
Encompass_Preferred
EPZ004777 is a potent, selective DOT1L inhibitor with IC50 of 0.4 nM in a cell-free assay and demonstrates >1,200-fold selectivity for DOT1L over all other tested PMTs. EPZ004777 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4676