Learn More
Apexbio Technology LLC EPZ004777 HCl, 5mg. Cas: 1380316-03-9 MFCD: MFCD27920717. DOT1L inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B15815
Encompass_Preferred
EPZ004777 is a potent and selective inhibitor of DOT1L, a protein methyltransferase catalyzing the methylation of lysine 4 of histone H3 (H3K4), with the half maximal inhibition concentration IC50 value of 400 pM [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5266