Learn More
Medchemexpress LLC Epz004777 hydrochloride | 1380316-03-9 | C28H42ClN7O4 | 5 MG
SDP
Catalog No. 5000372078
Encompass_Preferred
EPZ004777 hydrochloride is an effective and selective DOT1L inhibitor with an IC50 of 0.4 nM. It reduces the levels of H3K79me2 and H3K79me1, leading to a significant down-regulation of HOXA9 and MEIS1 expression.
  • Selectively inhibits the proliferation of MLL rearrangement cells
  • Promotes the differentiation and subsequent apoptosis of leukemia cells
  • Can be utilized for leukemia research
  • Potent, concentration-dependent inhibition of DOT1L enzyme activity with an IC50 of 400±100 pM
  • Remarkable selectivity for DOT1L over other HMTs
Catalog No. 50-003-72078 Supplier Medchemexpress LLC Supplier No. HY15227A5MG
EPZ004777 hydrochloride is an effective and selective DOT1L inhibitor with an IC50 of 0.4 nM. It reduces the levels of H3K79me2 and H3K79me1, leading to a significant down-regulation of HOXA9 and MEIS1 expression.
  • Selectively inhibits the proliferation of MLL rearrangement cells
  • Promotes the differentiation and subsequent apoptosis of leukemia cells
  • Can be utilized for leukemia research
  • Potent, concentration-dependent inhibition of DOT1L enzyme activity with an IC50 of 400±100 pM
  • Remarkable selectivity for DOT1L over other HMTs