Learn More
Medchemexpress LLC HY-15555 5mg Medchemexpress, EPZ005687 CAS:1396772-26-1 Purity:>98%
SDP
Catalog No. 502030647
Encompass_Preferred
Medchemexpress, HY-15555 5mg EPZ005687 CAS:1396772-26-1 EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0647 Supplier Medchemexpress LLC Supplier No. HY155555MG
Medchemexpress, HY-15555 5mg EPZ005687 CAS:1396772-26-1 EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.