Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EPZ011989 hydrochloride | 2095432-26-9 | 98.0% | 10 MG
SDP

Catalog No. 5000295226
Encompass_Preferred

EPZ011989 hydrochloride is the hydrochloride salt of a potent, orally active EZH2 inhibitor used as a research compound in epigenetics and oncology. It is intended for use in biochemical and cellular assays and for preclinical pharmacology studies requiring selective EZH2 modulation.

  • Potent EZH2 inhibition (Ki <3 nM).
  • High purity suitable for research use (98.0%).
  • Hydrochloride salt for improved solubility and handling.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Molecular weight 641.26 g/mol; formula C35H51N5O4·HCl.
  • Supplied as a small quantity vial (10 mg) for research applications.

Catalog No. 50-002-95226 Supplier Medchemexpress LLC Supplier No. HY16986B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EPZ011989 hydrochloride is the hydrochloride salt of a potent, orally active EZH2 inhibitor used as a research compound in epigenetics and oncology. It is intended for use in biochemical and cellular assays and for preclinical pharmacology studies requiring selective EZH2 modulation.

  • Potent EZH2 inhibition (Ki <3 nM).
  • High purity suitable for research use (98.0%).
  • Hydrochloride salt for improved solubility and handling.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Molecular weight 641.26 g/mol; formula C35H51N5O4·HCl.
  • Supplied as a small quantity vial (10 mg) for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.