Learn More
Medchemexpress LLC EPZ011989 (hydrochloride) | 2095432-26-9 | 99.4% | 642.27 | 25 MG
SDP
Catalog No. 5000394843
Encompass_Preferred
EPZ011989 hydrochloride is a potent and orally active Zeste Homolog 2 (EZH2) inhibitor. It demonstrates anti-tumor activity and functions as a click chemistry reagent. It is intended for research use only.
  • Potent and orally active EZH2 inhibitor
  • Ki value of <3 nM
  • Shows anti-tumor activity
  • Functions as a click chemistry reagent (contains an Alkyne group for CuAAc)
Catalog No. 50-003-94843 Supplier Medchemexpress LLC Supplier No. 25MGHY16986B
EPZ011989 hydrochloride is a potent and orally active Zeste Homolog 2 (EZH2) inhibitor. It demonstrates anti-tumor activity and functions as a click chemistry reagent. It is intended for research use only.
  • Potent and orally active EZH2 inhibitor
  • Ki value of <3 nM
  • Shows anti-tumor activity
  • Functions as a click chemistry reagent (contains an Alkyne group for CuAAc)