Learn More
Medchemexpress LLC HY-12727 10mg Medchemexpress, EPZ015666 CAS:1616391-65-1 Purity:>98%
SDP
Catalog No. 502028912
Encompass_Preferred
Medchemexpress, HY-12727 10mg EPZ015666 CAS:1616391-65-1 EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8912 Supplier Medchemexpress LLC Supplier No. HY1272710MG
Medchemexpress, HY-12727 10mg EPZ015666 CAS:1616391-65-1 EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.