Learn More
Selleck Chemical LLC EPZ015666
SDP
Catalog No. 5000000634
Encompass_Preferred
EPZ015666 (GSK3235025) is a potent selective and orally bioavailable PRMT5 inhibitor with Ki of 5 nM 20 000-fold selectivity over other PMTs
Catalog No. 50-000-00634 Supplier Selleck Chemical LLC Supplier No. S774825MG
EPZ015666 (GSK3235025) is a potent selective and orally bioavailable PRMT5 inhibitor with Ki of 5 nM 20 000-fold selectivity over other PMTs