Learn More
Apexbio Technology LLC EPZ015666 1616391-65-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062746
Encompass_Preferred
EPZ015666 (CAS 1616391-65-1) is a potent and selective inhibitor of protein arginine methyltransferase 5 (PRMT5) an enzyme implicated in several cellular pathways including tumorigenesis EPZ015666 inhibits PRMT5 with an IC50 of 22 nM and displays a Ki value of approximately 5 nM It exhibits over 20 000-fold selectivity for PRMT5 relative to other protein methyltransferases tested In mantle cell lymphoma (MCL) cell lines treatment with EPZ015666 reduced methylation levels of the PRMT5 substrate SmD3 and exerted concentration-dependent anti-proliferative effects Additionally EPZ015666 demonstrated dose-dependent tumor growth inhibition in mouse xenograft models of MCL highlighting its potential utility as a molecular tool in cancer biology research
Catalog No. 50-000-62746 Supplier Apexbio Technology LLC Supplier No. B49895.1
EPZ015666 (CAS 1616391-65-1) is a potent and selective inhibitor of protein arginine methyltransferase 5 (PRMT5) an enzyme implicated in several cellular pathways including tumorigenesis EPZ015666 inhibits PRMT5 with an IC50 of 22 nM and displays a Ki value of approximately 5 nM It exhibits over 20 000-fold selectivity for PRMT5 relative to other protein methyltransferases tested In mantle cell lymphoma (MCL) cell lines treatment with EPZ015666 reduced methylation levels of the PRMT5 substrate SmD3 and exerted concentration-dependent anti-proliferative effects Additionally EPZ015666 demonstrated dose-dependent tumor growth inhibition in mouse xenograft models of MCL highlighting its potential utility as a molecular tool in cancer biology research