Learn More
Apexbio Technology LLC EPZ015666, 25mg. Cas: 1616391-65-1 MFCD: MFCD28411588. PRMT5 inhibitor
SDP
Supplier:  Apexbio Technology LLC B498925
Encompass_Preferred
EPZ015666 is a potent and selective inhibitor of PRMT5 with IC50 value of 22 nM [1]. Protein arginine methyltransferase-5 (PRMT5) is an enzyme that plays an important role in many cellular processes including tumorigenesis [1]. EPZ015666 is an orally bioavailable PRMT5 inhibitor. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5366