Learn More
Selleck Chemical LLC EPZ015666 (GSK3235025) 5mg 1616391-65-1
SDP
Supplier:  Selleck Chemical LLC S77485MG
Encompass_Preferred
EPZ015666 (GSK3235025) is a potent, selective and orally bioavailable PRMT5 inhibitor with Ki of 5 nM, >20,000-fold selectivity over other PMTs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1229404