Learn More
Selleck Chemical LLC EPZ020411 2HCl
SDP
Supplier:  Selleck Chemical LLC S78205MG
Encompass_Preferred
EPZ020411 is a potent and selective small molecule PRMT6 inhibitor with an IC50 of 10 nM
Catalog No. 50-000-01588