Learn More
Medchemexpress LLC Epz020411 (hydrochloride) | 2070015-25-5 | 98.15% | 10MM 1ML
SDP
Catalog No. 5000340112
Encompass_Preferred
EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM. It demonstrates more than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8 and can be used for cancer research.
  • Selective inhibitor of PRMT6 (IC50 of 0.01 μM).
  • Exhibits over 10-fold selectivity for PRMT6 compared to PRMT1 and PRMT8.
  • Decreases H3R2 methylation in A375 cells.
  • Reduces neomycin- and cisplatin-induced cell apoptosis and increases hair cell survival.
  • Attenuates neomycin- and cisplatin-induced hearing loss in mice.
Catalog No. 50-003-40112 Supplier Medchemexpress LLC Supplier No. HY12970A10MM/1ML
EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM. It demonstrates more than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8 and can be used for cancer research.
  • Selective inhibitor of PRMT6 (IC50 of 0.01 μM).
  • Exhibits over 10-fold selectivity for PRMT6 compared to PRMT1 and PRMT8.
  • Decreases H3R2 methylation in A375 cells.
  • Reduces neomycin- and cisplatin-induced cell apoptosis and increases hair cell survival.
  • Attenuates neomycin- and cisplatin-induced hearing loss in mice.