Learn More
Medchemexpress LLC Epz020411 (hydrochloride) | 2070015-25-5 | 98.15% | 479.06 | 10 MG
SDP
Catalog No. 5000365154
Encompass_Preferred
EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM. It demonstrates more than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8. This compound can be utilized for cancer research.
  • Selective inhibitor of PRMT6
  • IC50 of 10 nM for PRMT6
  • Exhibits greater than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8
  • Suitable for cancer research
  • Off-white to light yellow solid appearance
  • Soluble in DMSO, 0.1 M HCl, and water
  • Stock solutions stable for 1 year at -80°C or 6 months at -20°C (under nitrogen)
  • Shows synergistic anti-proliferative effects on HCT116 and SW620 cells
Catalog No. 50-003-65154 Supplier Medchemexpress LLC Supplier No. 10MGHY12970A
EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM. It demonstrates more than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8. This compound can be utilized for cancer research.
  • Selective inhibitor of PRMT6
  • IC50 of 10 nM for PRMT6
  • Exhibits greater than 10-fold selectivity for PRMT6 over PRMT1 and PRMT8
  • Suitable for cancer research
  • Off-white to light yellow solid appearance
  • Soluble in DMSO, 0.1 M HCl, and water
  • Stock solutions stable for 1 year at -80°C or 6 months at -20°C (under nitrogen)
  • Shows synergistic anti-proliferative effects on HCT116 and SW620 cells