Learn More
TARGETMOL CHEMICALS INC EPZ020411 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580962
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EPZ020411 hydrochloride is a selective and potent small molecule PRMT6 inhibitor with an IC50 value of 10 nM. purity: 100%
Catalog No. 50-258-0962 Supplier TARGETMOL CHEMICALS INC Supplier No. T223255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EPZ020411 hydrochloride is a selective and potent small molecule PRMT6 inhibitor with an IC50 value of 10 nM. purity: 100%