Learn More
Epigentek Inc EpiQuik Global Histone H4 Acetylation Assay Kit (96 assays)
SDP
Catalog No. 501096347
Encompass_Preferred
The EpiQuik Global Histone H4 Acetylation Assay Kit is a convenient package of tools that allows the experimenter to measure global acetylation of histone H4 at tremendously fast speeds and consistency, superior and safer than all other current methods. The kit is ready-to-use and provides all the essential components needed to carry out a successful assay experiment. The EpiQuik Global Histone H4 Acetylation Assay Kit is suitable for specifically measuring global histone H4 acetylation using a variety of mammalian cells including fresh and frozen tissues, and cultured adherent and suspension cells.
Catalog No. 50-109-6347 Supplier Epigentek Inc Supplier No. P400996
The EpiQuik Global Histone H4 Acetylation Assay Kit is a convenient package of tools that allows the experimenter to measure global acetylation of histone H4 at tremendously fast speeds and consistency, superior and safer than all other current methods. The kit is ready-to-use and provides all the essential components needed to carry out a successful assay experiment. The EpiQuik Global Histone H4 Acetylation Assay Kit is suitable for specifically measuring global histone H4 acetylation using a variety of mammalian cells including fresh and frozen tissues, and cultured adherent and suspension cells.