Learn More
Epigentek Inc EpiQuik Global Histone H3K27 Tri-Methylation Assay Kit (96 assays)
SDP
Catalog No. 501096211
Encompass_Preferred
The EpiQuik Global Histone H3-K27 Tri-Methylation Assay Kit is suitable for specifically measuring global histone H3-K27 tri-methylation using a variety of mammalian cells including fresh and frozen tissues, cultured adherent and suspension cells.
Catalog No. 50-109-6211 Supplier Epigentek Inc Supplier No. P3020T96
The EpiQuik Global Histone H3-K27 Tri-Methylation Assay Kit is suitable for specifically measuring global histone H3-K27 tri-methylation using a variety of mammalian cells including fresh and frozen tissues, cultured adherent and suspension cells.