Learn More
Epigentek Inc EpiQuik Superoxide Dismutase Activity/Inhibition Assay Kit (Colorimetric) (96 assays)
SDP
Catalog No. 501096057
Encompass_Preferred
This kit utilizes a dye that produces a water-soluble formazan upon reduction with superoxide anion. The rate of the reduction with a superoxide anion is linearly related to the xanthine oxidase (XO) activity, and is inhibited by SOD. Therefore, the inhibition activity of SOD can be determined by measuring superoxide anion-caused formation of water-soluble dye (color intensity). The higher the SOD activity, the less the formation of formazan and the lower the OD value.
Catalog No. 50-109-6057 Supplier Epigentek Inc Supplier No. OP00011
This kit utilizes a dye that produces a water-soluble formazan upon reduction with superoxide anion. The rate of the reduction with a superoxide anion is linearly related to the xanthine oxidase (XO) activity, and is inhibited by SOD. Therefore, the inhibition activity of SOD can be determined by measuring superoxide anion-caused formation of water-soluble dye (color intensity). The higher the SOD activity, the less the formation of formazan and the lower the OD value.