Learn More
Epigentek Inc EpiQuik Total Histone Extraction HT Kit (192 extractions)
SDP
Catalog No. 501096067
Encompass_Preferred
The EpiQuik Total Histone Extraction HT Kit provides a simple and high throughput microplate method for extracting histone proteins used for a variety of applications, which include histone modifications such as acetylation, methylation, and sumoylation. This kit is also specifically designed to meet the requirements of histone extracts used in EpiQuik histone quantification assays. The kit can be used to directly extract histones from mammalian cells cultured in a 96-well plate. The EpiQuik Total Histone Extraction HT Kit has the fastest procedure available on the market, allowing completion within 60 minutes.
Catalog No. 50-109-6067 Supplier Epigentek Inc Supplier No. OP0007192
The EpiQuik Total Histone Extraction HT Kit provides a simple and high throughput microplate method for extracting histone proteins used for a variety of applications, which include histone modifications such as acetylation, methylation, and sumoylation. This kit is also specifically designed to meet the requirements of histone extracts used in EpiQuik histone quantification assays. The kit can be used to directly extract histones from mammalian cells cultured in a 96-well plate. The EpiQuik Total Histone Extraction HT Kit has the fastest procedure available on the market, allowing completion within 60 minutes.