Learn More
Enzo Life Sciences EQ1 NHS ester (1mg)
SDP
Catalog No. 502013473
Encompass_Preferred
EQ1 NHS ester is an amine-reactive quencher dye. It has a broad visible absorption but no detectable fluorescence emission, making it useful as an acceptor in fluorescence  resonance energy transfer (FRET) applications. It can be used to quench reporter dyes: FAM, Cy2, TET, JOE, VIC, HEX, Cy3, NED, TAMRA, Cy3.5, ROX, Texas Red, Quasar 570, CAL Fluor Gold 540, CAL Fluor Orange 560, CAL Fluor Red 590, CAL Fluor Red 610, Alexa Fluor 430, Alexa Fluor 488, Alexa Fluor 532, Alexa Fluor 546, Alexa Fluor 555, Alexa Fluor 568, Alexa Fluor 594, DyLight 488, DyLight 550, DyLight 594, ATTO 488, ATTO 520, ATTO 532, ATTO Rho6G, ATTO 550, ATTO 565, ATTO Rho11, ATTO 590, ATTO 594. Purity: ≥80% (HPLC). Solubility: Soluble in DMSO or DMF. Long Term Storage: -20°C. Applications: Fluorescent detection FRET.
Catalog No. 50-201-3473 Supplier Enzo Life Sciences Supplier No. ENZCHM1650001
EQ1 NHS ester is an amine-reactive quencher dye. It has a broad visible absorption but no detectable fluorescence emission, making it useful as an acceptor in fluorescence  resonance energy transfer (FRET) applications. It can be used to quench reporter dyes: FAM, Cy2, TET, JOE, VIC, HEX, Cy3, NED, TAMRA, Cy3.5, ROX, Texas Red, Quasar 570, CAL Fluor Gold 540, CAL Fluor Orange 560, CAL Fluor Red 590, CAL Fluor Red 610, Alexa Fluor 430, Alexa Fluor 488, Alexa Fluor 532, Alexa Fluor 546, Alexa Fluor 555, Alexa Fluor 568, Alexa Fluor 594, DyLight 488, DyLight 550, DyLight 594, ATTO 488, ATTO 520, ATTO 532, ATTO Rho6G, ATTO 550, ATTO 565, ATTO Rho11, ATTO 590, ATTO 594. Purity: ≥80% (HPLC). Solubility: Soluble in DMSO or DMF. Long Term Storage: -20°C. Applications: Fluorescent detection FRET.