Learn More
SOMNI Scientific™ Equaflow 10XL Mouse/Sm Rat Manifold
SDP
Catalog No. 04777135 Shop All Somni Scientific Products
Change view
Click to view available options
Model:
Equaflow 10XL Mouse/Sm Rat Manifold
Catalog No. Model
04-777-135 Equaflow 10XL Mouse/Sm Rat Manifold
1 options
Catalog No. 04-777-135 Supplier SOMNI Scientific™ Supplier No. EM16011
Add to Cart
Add to Cart
Request a Quote
Provides state of the art anesthetic gas delivery and scavenging just like the Equaflow 3. The difference is, the 10XL has greater spacing in-between nose cones for larger rodents.
  • Safeguards researchers from exposure
  • Designed for use with SOMNI™ EPS-3 or attenuated in-house vacuum system
  • Meets NIOSH standards for waste gas safety, when used properly
  • Hyper branched equal anesthetic gas delivery to each nosecone prevents animal awakening
  • Hyper branched active scavenging to each nosecone assuring user safety and compliance
  • Animals lay in a natural position during anesthesia (no head torqueing)
  • Non-auto fluorescent/ luminescent material
  • Optical imaging compatible (from above or below)
  • Integrated magnets assure consistent placement of animal and manifold
  • Includes positioners and base plate
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia, Imaging
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15001, WG-15101, OC-21003
Material Plastic
Length (Metric) 26 cm
Width (Metric) 5 cm
Model Equaflow 10XL Mouse/Sm Rat Manifold
Includes 12 x 12 plate, dividers, color coded tubing with adapters
For Veterinary Use Only