Learn More
SOMNI Scientific™ Equaflow 5 Mouse Manifold
SDP
Catalog No. 04777131 Shop All Somni Scientific Products
Change view
Click to view available options
Model:
Equaflow 5 Mouse Manifold
Catalog No. Model
04-777-131 Equaflow 5 Mouse Manifold
1 options
Catalog No. 04-777-131 Supplier SOMNI Scientific™ Supplier No. EM16005
Add to Cart
Add to Cart
Provides state of the art hyperbranched anesthetic gas delivery and scavenging equally to each nose cone
  • Meets NIOSH standards for waste gas safety, when used properly
  • Hyper branched equal anesthetic gas delivery to each nosecone prevents animal awakening
  • Hyper branched active scavenging to each nosecone assuring user safety and compliance
  • Animals lay in a natural position during anesthesia (no head torqueing)
  • Non-auto fluorescent/ luminescent material
  • Optical imaging compatible (from above or below)
  • Integrated magnets assure consistent placement of animal and manifold
  • Includes positioners and base plate
  • Made for use with SOMNI™ EPS-3 or attenuated vacuum system
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia, Imaging
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15001, WG-15101, OC-21003
Material Plastic
Length (Metric) 19.8 cm
Width (Metric) 3.2 cm
Model Equaflow 5 Mouse Manifold
Includes 12 x 12 plate, dividers, color coded tubing with adapters
For Veterinary Use Only