Learn More
SOMNI Scientific™ Equaflow Large Rat Manifold
SDP
Catalog No. 04777133 Shop All Somni Scientific Products
Change view
Click to view available options
Model:
Equaflow Large Rat Manifold
Catalog No. Model
04-777-133 Equaflow Large Rat Manifold
1 options
Catalog No. 04-777-133 Supplier SOMNI Scientific™ Supplier No. EM16007
Add to Cart
Add to Cart
Provides state of the art anesthesia delivery with scavenging built right in. This feature safeguards researchers and technicians from exposure during procedures.
  • Anesthesia delivery with built in scavenging
  • Anesthetic stays within the nosecone, when used properly
  • Meets NIOSH standards for waste gas safety
  • For use with SOMNI™ EPS-3 or attenuated in-house vacuum
  • Animals lay in a flat, natural position during anesthesia
  • Magnetic placement and positioners included
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia, Imaging
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15001, WG-15101, OC-21003
Material Plastic
Length (Metric) 8.3 cm
Width (Metric) 5.6 cm
Model Equaflow Large Rat Manifold
Includes 12 x 12 plate, magnetic positioners, color coded tubing with adapters
For Veterinary Use Only