Learn More
VAZYME INTERNATIONAL LLC Vazyme Equalbit 1 × dsDNA HS Assay Kit, 100 Reactions, 0.2 to 100 ng dsDNA
Catalog No. NC3979710
Encompass
The Equalbit 1 × dsDNA HS Assay Kit is a simple, sensitive, and accurate fluorescence quantitative detection kit for double-stranded DNA (dsDNA). It features a pre-mixed working solution with fluorescent dye and dsDNA standards, offering linearity from 0.2 to 100 ng dsDNA for precise quantification (10 pg/μl to 100 ng/μl). It tolerates common contaminants, enabling straightforward dsDNA quantification with a fluorescence quantifier.
  • Simple operation due to ready-to-use pre-mix, no working solution preparation needed.
  • Accurately quantifies 0.2 to 100 ng of dsDNA.
  • Specifically detects dsDNA and tolerates common contaminants.
  • Rapid dye binding, achieving saturation within 2 minutes.
  • High stability from strictly controlled standard production.
  • Easy to operate with a Qubit fluorometer.
Catalog No. NC3979710 Supplier VAZYME INTERNATIONAL LLC Supplier No. EQ12101
The Equalbit 1 × dsDNA HS Assay Kit is a simple, sensitive, and accurate fluorescence quantitative detection kit for double-stranded DNA (dsDNA). It features a pre-mixed working solution with fluorescent dye and dsDNA standards, offering linearity from 0.2 to 100 ng dsDNA for precise quantification (10 pg/μl to 100 ng/μl). It tolerates common contaminants, enabling straightforward dsDNA quantification with a fluorescence quantifier.
  • Simple operation due to ready-to-use pre-mix, no working solution preparation needed.
  • Accurately quantifies 0.2 to 100 ng of dsDNA.
  • Specifically detects dsDNA and tolerates common contaminants.
  • Rapid dye binding, achieving saturation within 2 minutes.
  • High stability from strictly controlled standard production.
  • Easy to operate with a Qubit fluorometer.