Learn More
Analytical Sales & Services EquaVAP 24-Well Evaporator, 18mm
Catalog No. 5000804804
Encompass
EquaVAP 24-Well Blowdown Evaporator, 18mm \n\n• 18mm center-to-center needle distance\n• Use with 24-well polypropylene collection plates (part numbers 24108 or 24727)
Catalog No. 50-008-04804 Supplier Analytical Sales & Services Supplier No. 2302418
EquaVAP 24-Well Blowdown Evaporator, 18mm \n\n• 18mm center-to-center needle distance\n• Use with 24-well polypropylene collection plates (part numbers 24108 or 24727)