Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Equi-Stat Antibacterial Handwash
SDP

Catalog No. 21033255
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-255 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-255 Supplier StatLab Supplier No. 104872
Add to Cart
Add to Cart

Antibacterial hand wash containing triclosan

  • Contains Triclosan, lanolin and aloe vera
  • Triclosan bonds to skin for persistence against incubation under gloves
  • 18 oz./540 mL dispenser bottle

Specifications

Product Type Anti-bacterial Hand Soap
Color Blue
Volume (English) 18 oz.
Volume (Metric) 540 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.