Learn More
Med Vet International Equidone Oral Gel (Domperidone)
Catalog No. 502828082
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Equidone Gel is indicated for the prevention of fescue toxicosis in periparturient mares. This disposable 25cc multi-dose syringe contains 2.75 g (110 mg/cc) of domperidone suspended in an oral gel and is to be administered at a dosage of 0.5mglb (1.1mg/kg) once daily starting 10 to 15 days prior to Expected Foaling Date (EFD). This treatment may be continued for up to 5 days, after foaling, if mares are not producing adequate milk following foaling.
Catalog No. 50-282-8082 Supplier Med Vet International Supplier No. RXEQUIGEL
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Equidone Gel is indicated for the prevention of fescue toxicosis in periparturient mares. This disposable 25cc multi-dose syringe contains 2.75 g (110 mg/cc) of domperidone suspended in an oral gel and is to be administered at a dosage of 0.5mglb (1.1mg/kg) once daily starting 10 to 15 days prior to Expected Foaling Date (EFD). This treatment may be continued for up to 5 days, after foaling, if mares are not producing adequate milk following foaling.