Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Equilateral Prism
GREENER_CHOICESCI_ED

Catalog No. S160131 Shop All Supertek Scientific Products
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S160131 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160131 Supplier Supertek™ Supplier No. PH30305A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate refraction principles and light physics, using this Supertek™ Crystal Clear Acrylic, Equilateral Prism.

  • Crystal-Clear Acrylic
  • 1 x 4 in. desirable size

Specifications

Grade 1 to 14
Includes Equilateral Prism
Material Acrylic
Class Size 1 Student
Product Type Prism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.