Learn More
Medchemexpress LLC Equilin 3-sulfate-d4 (sodium) | 285979-81-9 | 95.0% | 375.4 g/mol | C18H16D4NaO5S | 5 MG
SDP
Catalog No. 5000262642
Encompass_Preferred
Equilin 3-Sulfate-d4 (sodium) is the deuterium-labeled sodium salt of equilin 3-sulfate supplied for analytical and research use. It is intended as a stable-isotope internal standard or tracer for quantitative mass spectrometry and NMR applications. The material is typically provided as a solid and should be stored sealed at -20 °C; in solvent it is stable at -80 °C for up to 6 months or -20 °C for 1 month.
Catalog No. 50-002-62642 Supplier Medchemexpress LLC Supplier No. HY132641S5MG
Equilin 3-Sulfate-d4 (sodium) is the deuterium-labeled sodium salt of equilin 3-sulfate supplied for analytical and research use. It is intended as a stable-isotope internal standard or tracer for quantitative mass spectrometry and NMR applications. The material is typically provided as a solid and should be stored sealed at -20 °C; in solvent it is stable at -80 °C for up to 6 months or -20 °C for 1 month.