Learn More
Creative Biomart Equine anti-thymocyte globulin/Antilymphocyte immunoglobulin (horse)
Supplier:  Creative Biomart THP0107
Encompass_Preferred
Equine anti-thymocyte globulin is composed of purified gamma globulin containing primarily IgG against human thymus lymphocytes. It is formed by inoculating a horse with an antigen (human thymoyctes) which then induces the horse immune system's B-lymphocytes to produce IgG immunoglobulins specific for that antigen. The result is polyclonal IgG that is then purified from the horse's serum to produce a usable product that can be used for immunosuppression.
Catalog No. 50-208-4158