Learn More
Kingfisher Biotech Inc Equine CCL3 (MIP-1 alpha) Polyclonal Antibody - Biotinylated (50 ug)
SDP
Supplier:  Kingfisher Biotech Inc PBB0401E050
Encompass
The Equine CCL3 (MIP-1 alpha) polyclonal antibody is labeled and has been qualified for use in ELISA. It is the detection antibody in the Kingfisher Biotech Equine CCL3 ELISA. The Equine CCL3 polyclonal antibody was produced in rabbits and is antigen-affinity purified. For research use only. Made in the USA
Catalog No. 50-233-2236