Learn More
Med Vet International Equine Dental Fulcrum
Catalog No. 5000584210
Encompass_Preferred
Use as a fulcrum to gain leverage in conjunction with a molar extraction forceps when extracting large cheek teeth. Dual End. 22" (54cm) L. Limited Supply. No backorders. No Returns
Catalog No. 50-005-84210 Supplier Med Vet International Supplier No. XJOR705F
Use as a fulcrum to gain leverage in conjunction with a molar extraction forceps when extracting large cheek teeth. Dual End. 22" (54cm) L. Limited Supply. No backorders. No Returns