Learn More
Med Vet International Equine Dental Instruments, Dental Probe Heavy Tip, 19in, J0441, Each
Catalog No. 5000584449
Encompass_Preferred
A large dental patient like a horse needs oversized equipment. These dental probes, scalers, and explorers are giant sized to 19" with knurled handles. If you do a lot of equine dentistry, you are going to find them very useful. Calibrated probe is used to measure depth of periodontal pockets
Catalog No. 50-005-84449 Supplier Med Vet International Supplier No. JOR441
A large dental patient like a horse needs oversized equipment. These dental probes, scalers, and explorers are giant sized to 19" with knurled handles. If you do a lot of equine dentistry, you are going to find them very useful. Calibrated probe is used to measure depth of periodontal pockets