Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Equine Donor Serum

Catalog No. ICN29211Z0 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
Color:
Golden Brown
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
ICN29211Z0 - -
ICN2921154 500 mL Golden Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN29211Z0 Supplier MP Biomedicals™ Supplier No. 29211Z0

missing translation for 'validMainframeMsg'

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.