Learn More
Kingfisher Biotech Inc Equine GM-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0022E025
Encompass
The Equine GM-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine GM-CSF applications are for cell culture, ELISA standard, and Western Blot Control. The Equine GM-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Equine GM-CSF Specifications: (Molecular Weight: 15.2 kDa) (Amino Acid Sequence: APTRQPSPVT RPWQHVDAIK EALSLLNNSS DTAAIMNETV EVVSETFDAE ELTCLQTRLK LYKQGLRGSL IKLEGPLTMM ASHYKQHCPP TLETSCATQM ITFKSFKKNL KDFLFEIPFD CWSQPRSKAG LPARS) (Gene ID: 100033981). For research use only. Made in the USA
Catalog No. 50-241-3455