Learn More
Kingfisher Biotech Inc Equine IFN gamma Polyclonal Antibody - Biotinylated (50 ug)
SDP
Supplier:  Kingfisher Biotech Inc PBB0282E050
Encompass
The Equine IFN gamma polyclonal antibody is labeled and has been qualified for use in ELISA. It is the detection antibody in the Kingfisher Biotech Equine IFN Gamma ELISA. The Equine IFN gamma polyclonal antibody was produced in rabits and is antigen-affinity purified. The reactivity by species is: (Bovine IFN-gamma - Weak) (Canine IFN-gamma - Moderate) (Chicken IFN-gamma - None) (Dolphin IFN-gamma - Moderate) (Equine IFN-gamma - Strong) (Feline IFN-gamma - Weak) (Human IFN-gamma - Moderate) (Mouse IFN-gamma - Weak) (Rabbit IFN-gamma - Weak) (Swine IFN-gamma - Weak). For research use only. Made in the USA
Catalog No. 50-233-2443