Learn More
Kingfisher Biotech Inc Equine IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0012E005
Encompass
The Equine IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Equine IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Equine IFN gamma Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: YYCQAAFFKE IENLKEYFNA SNPDVGDGGP LFLDILKNWK EDSDKKIIQS QIVSFYFKLF ENLKDNQVIQ KSMDTIKEDL FAKFFNSSTS KLEDFQKLIQ IPVNDLKVQR KAISELIKVM NDLSPKANLR KRKRSQNPFR GRRALQ (146)) (Gene ID: 100034181). For research use only. Made in the USA
Catalog No. 50-241-3861