Learn More
Raybiotech Inc Equine IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELEIFNG1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.13 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kits detects equine IFN-gamma. This antibody pair has less than 20% cross-reactivity with recombinant canine IFN-gamma is observed, less than 6% cross-reactivity with recombinant feline IFN-gamma and recombinant bovine IFN-gamma is observed, and less than 0.2% cross-reactivity with recombinant human IFN-gamma, recombinant mouse IFN-gamma, recombinant rat IFN-gamma, recombinant porcine IFN-gamma, recombinant rhesus macaque IFN-gamma, and recombinant cotton rat IFN-gamma is observed.
Catalog No. 50-194-8903