Learn More
Kingfisher Biotech Inc Equine IL-10 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0757E005
Encompass
The Equine IL-10 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine IL-10 applications are for cell culture, ELISA standard, and Western Blot Control. The Equine IL-10 yeast-derived recombinant protein can be purchased in multiple sizes. Equine IL-10 Specifications: (Molecular Weight: 18.6 kDa) (Amino Acid Sequence: SRDRGTQSEN SCTHFPTSLP HMLHELRAAF SRVKTFFQMK DQLDNMLLNG SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRVRLRRCHR FLPCENKSKA VEQVKSAFSK LQEKGVYKAM SEFDIFINYI EAYMTTKMKN (170)) (Gene ID: 100034187). For research use only. Made in the USA
Catalog No. 50-241-3321