Learn More
Kingfisher Biotech Inc Swine IL-28B ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY0703E003
Encompass_Preferred
The Swine IL-28B (IFN Lambda) ELISA contains capture antibody, standard, and detection antibody for development of a Swine IL-28B ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include Interleukin 28B, IFNL3. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5189